missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3071-100
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Erythropoietin R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2057 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 38407 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P19235 | |
| EPOR | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
| 100 μg | |
| Primary | |
| Detects human Erythropoietin Receptor (Epo R) in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Epo, recombinant mouse Epo R, or rhThrombopoietin R was observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction