missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CPT1A Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
234.00 € - 537.00 €
Specifications
| Antigen | CPT1A |
|---|---|
| Clone | 1081618 |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30241010
|
R&D Systems
MAB11570-SP |
25 μg |
234.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30241014
|
R&D Systems
MAB11570-100 |
100 μg |
537.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPT1A Monoclonal antibody specifically detects CPT1A in Human samples. It is validated for ImmunohistochemistrySpecifications
| CPT1A | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Lyophilized | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, Carnitine palmitoyltransferase 1A, carnitine palmitoyltransferase 1A (liver), carnitine palmitoyltransferase I, liver, CPT I, CPT1, CPT1-L, CPT1-LEC 2.3.1.21, CPTI-L, EC 2.3.1, EC 2.3.1.21, L-CPT1 | |
| IgG2b | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1081618 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P50416 | |
| 1374 | |
| Primary | |
| Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title