missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CPT1A Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11570-100
This item is not returnable.
View return policy
Description
CPT1A Monoclonal antibody specifically detects CPT1A in Human samples. It is validated for Immunohistochemistry
Specifications
| CPT1A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, Carnitine palmitoyltransferase 1A, carnitine palmitoyltransferase 1A (liver), carnitine palmitoyltransferase I, liver, CPT I, CPT1, CPT1-L, CPT1-LEC 2.3.1.21, CPTI-L, EC 2.3.1, EC 2.3.1.21, L-CPT1 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| IgG2b |
| Immunohistochemistry | |
| 1081618 | |
| Immunohistochemistry 3-25 μg/mL | |
| P50416 | |
| Mouse | |
| RUO | |
| 1374 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction