missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C3a Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
203.00 € - 433.00 €
Specifications
| Antigen | Complement Component C3a |
|---|---|
| Clone | 354113 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733539
![]() |
R&D Systems
MAB3677-SP |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18199530
|
R&D Systems
MAB3677 |
100 μg |
433.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C3a Monoclonal specifically detects Complement Component C3a in Human samples. It is validated for Western Blot.Specifications
| Complement Component C3a | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01024 | |
| C3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 354113 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 718 | |
| E. coli-derived recombinant human Complement Component C3a Ser672-Arg748 Accession # P01024 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Component C3a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Complement Component C5a or rhComplement Component C3d is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title