missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C3a Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3677-SP
This item is not returnable.
View return policy
Description
Complement Component C3a Monoclonal specifically detects Complement Component C3a in Human samples. It is validated for Western Blot.
Specifications
| Complement Component C3a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P01024 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 718 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 354113 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C3 | |
| E. coli-derived recombinant human Complement Component C3a Ser672-Arg748 Accession # P01024 | |
| 25 μg | |
| Primary | |
| Detects human Complement Component C3a in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Complement Component C5a or rhComplement Component C3d is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction