missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Choline Acetyltransferase/ChAT Antibody, R&D Systems™

Mouse Monoclonal Antibody
214.00 € - 461.00 €
Specifications
| Antigen | Choline Acetyltransferase/ChAT |
|---|---|
| Clone | 334008 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719507
![]() |
R&D Systems
MAB3447-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18102971
![]() |
R&D Systems
MAB3447 |
100 μg |
461.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Choline Acetyltransferase/ChAT Monoclonal specifically detects Choline Acetyltransferase/ChAT in Human samples. It is validated for Western Blot.Specifications
| Choline Acetyltransferase/ChAT | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_066266 | |
| 1103 | |
| E. coli-derived recombinant human Choline Acetyltransferase/ChAT isoform 1 Ala2-Pro630 Accession # NP_066266 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Choline Acetyltransferase/ChAT in direct ELISAs and Western blots. Recognizes both ChAT isoform 1 (ChAT-69) and isoform 2 (ChAT-82). |
| 334008 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTASE, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
| CHAT | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title