missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Choline Acetyltransferase/ChAT Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3447-SP
This item is not returnable.
View return policy
Description
Choline Acetyltransferase/ChAT Monoclonal specifically detects Choline Acetyltransferase/ChAT in Human samples. It is validated for Western Blot.
Specifications
| Choline Acetyltransferase/ChAT | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_066266 | |
| CHAT | |
| E. coli-derived recombinant human Choline Acetyltransferase/ChAT isoform 1 Ala2-Pro630 Accession # NP_066266 | |
| 25 μg | |
| Primary | |
| Detects human Choline Acetyltransferase/ChAT in direct ELISAs and Western blots. Recognizes both ChAT isoform 1 (ChAT-69) and isoform 2 (ChAT-82). | |
| Human | |
| Purified |
| Western Blot | |
| 334008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTASE, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1103 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction