missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BMPR2 Partial ORF (NP_001195.2, 939 a.a. - 1037 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000659-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16053685

  • 349.00 € / 10µg
Estimated Shipment: 30-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs, which are members of the TGF-beta superfamily. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in this gene have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease. [provided by RefSeq]

Sequence: NSLDLSATNVLDGSSIQIGESTQDGKSGSGEKIKKRVKTPYSLKRWRPSTWVISTESLDCEVNNNGSNRAVHSKSSTAVYLAEGGTATTMVSKDIGMNC
Specifications

Specifications

NP_001195.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NSLDLSATNVLDGSSIQIGESTQDGKSGSGEKIKKRVKTPYSLKRWRPSTWVISTESLDCEVNNNGSNRAVHSKSSTAVYLAEGGTATTMVSKDIGMNC
RUO
BMPR2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
659
BMPR2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BMPR-II/BMPR3/BMR2/BRK-3/FLJ41585/FLJ76945/PPH1/T-ALK
BMPR2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions