Learn More
Abnova™ Human BMPR2 Partial ORF (NP_001195.2, 939 a.a. - 1037 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_001195.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 659 |
Molecular Weight (g/mol) | 36.63kDa |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16063685
|
Abnova™
H00000659-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053685
|
Abnova™
H00000659-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs, which are members of the TGF-beta superfamily. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in this gene have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease. [provided by RefSeq]
Sequence: NSLDLSATNVLDGSSIQIGESTQDGKSGSGEKIKKRVKTPYSLKRWRPSTWVISTESLDCEVNNNGSNRAVHSKSSTAVYLAEGGTATTMVSKDIGMNCSpécification
NP_001195.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMPR-II/BMPR3/BMR2/BRK-3/FLJ41585/FLJ76945/PPH1/T-ALK | |
BMPR2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
659 | |
BMPR2 (Human) Recombinant Protein (Q01) | |
NSLDLSATNVLDGSSIQIGESTQDGKSGSGEKIKKRVKTPYSLKRWRPSTWVISTESLDCEVNNNGSNRAVHSKSSTAVYLAEGGTATTMVSKDIGMNC | |
RUO | |
BMPR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |