missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ASB8 Partial ORF (NP_077000, 179 a.a. - 288 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_077000 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 140461 |
Molecular Weight (g/mol) | 37.84kDa |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
16181917
|
Abnova™
H00140461-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171917
|
Abnova™
H00140461-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Sequence: VINLIGQTPISRLVALLVRGLGTEKEDSCFELLHRAVGHFELRKNGTMPREVARDPQLCEKLTVLCSAPGTLKTLARYAVRRSLGLQYLPDAVKGLPLPASLKEYLLLLESpezifikation
NP_077000 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ21255/MGC5540/PP14212 | |
ASB8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
140461 | |
ASB8 (Human) Recombinant Protein (Q01) | |
VINLIGQTPISRLVALLVRGLGTEKEDSCFELLHRAVGHFELRKNGTMPREVARDPQLCEKLTVLCSAPGTLKTLARYAVRRSLGLQYLPDAVKGLPLPASLKEYLLLLE | |
RUO | |
ASB8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |