missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASB8 Partial ORF (NP_077000, 179 a.a. - 288 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00140461-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16171917

  • 335.00 € / 10µg
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VINLIGQTPISRLVALLVRGLGTEKEDSCFELLHRAVGHFELRKNGTMPREVARDPQLCEKLTVLCSAPGTLKTLARYAVRRSLGLQYLPDAVKGLPLPASLKEYLLLLE
Specifications

Specifications

NP_077000
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VINLIGQTPISRLVALLVRGLGTEKEDSCFELLHRAVGHFELRKNGTMPREVARDPQLCEKLTVLCSAPGTLKTLARYAVRRSLGLQYLPDAVKGLPLPASLKEYLLLLE
RUO
ASB8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
140461
ASB8 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21255/MGC5540/PP14212
ASB8
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions