Learn More
Hichrom™ HPLC column EXSAX-5-250A SAX ion exchange stainless steel 5μm 250mm x 4.6mm Exsil
Brand: Hichrom™ EXSAX5250A
Additional Details : Weight : 1.00000kg
Brand: Hichrom™ EXSAX5250A
Additional Details : Weight : 1.00000kg