Learn More
HMN-154, MedChemExpress
Product Code. 16486487
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486487 5 mg 5mg
16484826 100 mg 100mg
16470147 50 mg 50mg
16443768 10 mg 10mg
16420989 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16486487 Supplier Medchem Express Supplier No. HY103001
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress HMN-154 is a novel benzenesulfonamide anticancer compound; inhibits KB and colon38 cells with IC50 values of 0.0026 and 0.003 μg/mL, respectively.
HMN-154 is a novel benzenesulfonamide anticancer compound
Specifications
Chemical Name or Material HMN-154
CAS 173528-92-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H18N2O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 15 mg/mL (40.94 mM)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=CC=CC=C2/C=C/C3=CC=NC=C3)=O
Molecular Weight (g/mol) 366.43
Formula Weight 366.43
Percent Purity 97.18%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death