missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 35 publications
267.00 € - 635.00 €
Specifications
| Antigen | Fibronectin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported by customer review |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18757773
|
Novus Biologicals
NBP1-91258SS |
0.025 mL |
267.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18786343
|
Novus Biologicals
NBP1-91258 |
0.1 mL |
635.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Fibronectin | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported by customer review | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| PBS, 30% Glycerol with 0.05% Sodium Azide | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| FN1 | |
| IgG | |
| Affinity Purified | |
| Human and Mouse. Based on immunogen's sequence homology, this antibody is predicted to react with Chicken (91%), Rat (95%), Canine/Dog (100%), Bovine (100%) and Equine/Horse (100%). Immunogen similarity with other species: Xenopus (86%) and Newt (82%). |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline | |
| P02751 | |
| 2335 | |
| A synthetic peptide made toward the C-terminal region of the human Fibronectin protein (within residues 2250-2300). [Swiss-Prot: P02751] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title