missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 35 publications
Brand: Novus Biologicals NBP1-91258SS
This item is not returnable.
View return policy
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Fibronectin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 30% Glycerol with 0.05% Sodium Azide | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and Mouse. Based on immunogen's sequence homology, this antibody is predicted to react with Chicken (91%), Rat (95%), Canine/Dog (100%), Bovine (100%) and Equine/Horse (100%). Immunogen similarity with other species: Xenopus (86%) and Newt (82%). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported by customer review | |
| P02751 | |
| FN1 | |
| A synthetic peptide made toward the C-terminal region of the human Fibronectin protein (within residues 2250-2300). [Swiss-Prot: P02751] | |
| 0.025 mL | |
| Cancer, Cellular Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2335 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction