Learn More
(Heptafluorobutyro)phenone, 99%
Product Code. 11313746
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11313746
Brand: Thermo Scientific Alfa Aesar L16777.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 559-91-1 | C10H5F7O | 274.138 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 559-91-1
Molecular Formula C10H5F7O
Molecular Weight (g/mol) 274.138
MDL Number MFCD00077552
InChI Key UKYZDGAVBNKBPQ-UHFFFAOYSA-N
Synonym heptafluoropropyl phenyl ketone, heptafluorobutyrophenone, phenyl heptafluoropropyl ketone, pubchem20274, heptafluorobutyro phenone, 1-heptafluorobutyryl benzene, heptafluoropropylphenylketone, 2,2,3,3,4,4,4-heptafluoro-1-phenyl-butan-1-one
PubChem CID 238424
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-phenylbutan-1-one
SMILES C1=CC=C(C=C1)C(=O)C(C(C(F)(F)F)(F)F)(F)F
Specifications
Density 1.473
Boiling Point 170°C to 172°C
Refractive Index 1.414
Quantity 25 g
Beilstein 2056234
Formula Weight 274.14
Percent Purity 99%
Chemical Name or Material (Heptafluorobutyro)phenone