accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 559-91-1

FFFFFFFO

CAS RN 559-91-1

IUPAC Name: 2,2,3,3,4,4,4-heptafluoro-1-phenylbutan-1-one
Synonyms: 2,2,3,3,4,4,4-heptafluoro-1-phenylbutan-1-one
Molecular Weight (g/mol): 274.14
Molecular Formula: C10H5F7O
InChi Key: UKYZDGAVBNKBPQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results