Learn More
HEPES, CellPURE™, Fisher BioReagents
Product Code. 10394870
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. 10394870
Brand: Fisher BioReagents BP2937100
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Absorbance 0.05 AU max. at 260nm, 0.05 AU max. at 280nm, 0.05 AU max. at 290nm
CAS 7365-45-9
Melting Point 234°C
Color White
Decomposition Information 238°C
pH 5.0 to 6.5
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥99.5 %
Infrared Spectrum Conforms to reference
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
Packaging Poly Bottle
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-Na2-(2-ethanesulfonic acid)
Solubility Information Soluble in water
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Formula Weight 238.3
Percent Purity ≥99.5%
Grade CellPURE
Chemical Name or Material HEPES, Cell culture tested