missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega HeLaScribe™ Nuclear Extract in vitro Transcription Grade
HeLaScribe Nuclear Extract, in vitro Transcription Grade.
POA
Specifications
Content And Storage | <-65°C |
---|---|
For Use With (Application) | Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites |
Product Type | HeLaScribe Nuclear Extract, Transcription Grade |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10574013
|
Promega
E309.1 |
40 |
POA
Pack of 100 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- Cell-Free Extract for in vitro Transcription of Eukaryotic Genes
- Component of the HeLaScribe(R) Nuclear Extract in vitro Transcription System
- Source of transcription factors, DNA-binding proteins and RNA processing components
Specifications
<-65°C | |
Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites | |
HeLaScribe Nuclear Extract, Transcription Grade |