missing translation for 'onlineSavingsMsg'
Learn More

Promega HeLaScribe™ Nuclear Extract in vitro Transcription Grade

Product Code. 10574013
Encompass
Change view
Click to view available options
No. of Reactions:
40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions
10574013 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10574013 Supplier Promega Supplier No. E309.1
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HeLaScribe Nuclear Extract, in vitro Transcription Grade.

  • Cell-Free Extract for in vitro Transcription of Eukaryotic Genes
  • Component of the HeLaScribe(R) Nuclear Extract in vitro Transcription System
  • Source of transcription factors, DNA-binding proteins and RNA processing components

Specifications

Content And Storage <-65°C
For Use With (Application) Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites
No. of Reactions 40
Product Type HeLaScribe Nuclear Extract, Transcription Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.