Learn More
α-HCH 100 μg/mL in Acetone, Dr. Ehrenstorfer
Product Code. 30309082
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309082 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309082 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010365AC
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material alpha-HCH
CAS 319-84-6
Analyte or Component Names alpha-HCH
Shipping Condition Room Temperature
Molecular Formula C6 H6 Cl6
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym (1α,2α,3β,4α,5β,6β)-1,2,3,4,5,6-Hexachlorocyclohexane, (±)-α-HCH, (±)-α-Hexachlorocyclohexane, α-1,2,3,4,5,6-Hexachlorocyclohexane, α-666, α-BHC, α-Benzenehexachloride, α-Benzohexachloride, α-HCH, α-Hexachloran, α-Hexachlorane, α-Hexachlorcyclohexane, α-Hexachlorocyclohexane, α-Lindane
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4-,5+,6+/m0/s1
SMILES Cl[C@@H]1[C@@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H](Cl)[C@H]1Cl
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
Solvent or Matrix Acetone
Molecular Weight (g/mol) 290.83
Quantity 1 mL
Formula Weight 287.8601 g/mol