missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GUCY1B3 (Human) Recombinant Protein
Human GUCY1B3 full-length ORF ( AAH47620, 1 a.a. - 619 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH47620 |
---|---|
Gene ID (Entrez) | 2983 |
Name | guanylate cyclase 1, soluble, beta 3 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185931
|
Abnova™
H00002983-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16195931
|
Abnova™
H00002983-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MYGFVNHALELLVIRNYGPEVWEDIKKEAQLDEEGQFLVRIIYDDSKTYDLVAAASKVLNLNAGEILQMFGKMFFVFCQESGYDTILRVLGSNVREFLQNLDALHDHLATIYPGMRAPSFRCTDAEKGKGLILHYYSEREGLQDIVIGIIKTVAQQIHGTEIDMKVIQQRNEECDHTQFLIEEKESKEEDFYEDLDRFEENGTQESRISPYTFCKAFPFHIIFDRDLVVTQCGNAIYRVLPQLQPGNCSLLSVFSLVRPHIDISFHGILSHINTVFVLRSKEGLLDVEKLECEDELTGTEISCLRLKGQMIYLPEADSILFLCSPSVMNLDDLTRRGLYLSDIPLHDATRDLVLLGEQFREEYKLTQELEILTDRLQLTLRALEDEKKKTDTLLYSVLPPSVANELRHKRPVPAKRYDNVTILFSGIVGFNAFCSKHASGEGAMKIVNLLNDLYTRFDTLTDSRKNPFVYKVETVGDKYMTVSGLPEPCIHHARSICHLALDMMEIAGQVQVDGESVQITIGIHTGEVVTGVIGQRMPRYCLFGNTVNLTSRTETTGEKGKINVSEYTYRCLMSPENSDPQFHLEHRGPVSMKGKKEPMQVWFLSRKNTGTEETKQDDD
Specifications
AAH47620 | |
guanylate cyclase 1, soluble, beta 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
GC-S-beta-1, GC-SB3, GUC1B3, GUCB3, GUCSB3, GUCY1B1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2983 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GUCY1B3 | |
GST |