missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GUCY1B3 (Human) Recombinant Protein
Human GUCY1B3 full-length ORF ( AAH47620, 1 a.a. - 619 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002983-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MYGFVNHALELLVIRNYGPEVWEDIKKEAQLDEEGQFLVRIIYDDSKTYDLVAAASKVLNLNAGEILQMFGKMFFVFCQESGYDTILRVLGSNVREFLQNLDALHDHLATIYPGMRAPSFRCTDAEKGKGLILHYYSEREGLQDIVIGIIKTVAQQIHGTEIDMKVIQQRNEECDHTQFLIEEKESKEEDFYEDLDRFEENGTQESRISPYTFCKAFPFHIIFDRDLVVTQCGNAIYRVLPQLQPGNCSLLSVFSLVRPHIDISFHGILSHINTVFVLRSKEGLLDVEKLECEDELTGTEISCLRLKGQMIYLPEADSILFLCSPSVMNLDDLTRRGLYLSDIPLHDATRDLVLLGEQFREEYKLTQELEILTDRLQLTLRALEDEKKKTDTLLYSVLPPSVANELRHKRPVPAKRYDNVTILFSGIVGFNAFCSKHASGEGAMKIVNLLNDLYTRFDTLTDSRKNPFVYKVETVGDKYMTVSGLPEPCIHHARSICHLALDMMEIAGQVQVDGESVQITIGIHTGEVVTGVIGQRMPRYCLFGNTVNLTSRTETTGEKGKINVSEYTYRCLMSPENSDPQFHLEHRGPVSMKGKKEPMQVWFLSRKNTGTEETKQDDD
Specifications
AAH47620 | |
guanylate cyclase 1, soluble, beta 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GUCY1B3 | |
GST |
2983 | |
Wheat germ expression system | |
25 μg | |
GC-S-beta-1, GC-SB3, GUC1B3, GUCB3, GUCSB3, GUCY1B1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |