missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grace™ Vydac™ C18 (218TP) HPLC Columns
Description
- Provide reliable, high resolution of proteins and large peptides
- Relied upon in numerous commercial scale protein purifications
- Vydac TP is completely scalable to ton quantities
- Threaded
Specifications
Specifications
| Diameter (Metric) | 10 mm |
| Flow Rate | 5 mL/min |
| For Use With | LC/MS Separation |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| Pore Size | 300 Å |
| Surface Area | 60 m²/g |
| USP Type | L1 |
| Stationary Phase | C18 |
| Packing Material | Silica |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?