missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Jones Genesis™ C18 Reversed Phase HPLC Columns

Product Code. 11370041
Encompass
Click to view available options
Length (Metric):
100 mm
Diameter (Metric):
4.6 mm
Particle Size:
4 μm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11370041

Brand: Grace™ FM10960E

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

A unique proprietary end-capping reagent provides freedom from residual silanols and enhanced stability under low-pH operating conditions

  • Excellent peak symmetry
  • Exceptional stability from 1 to 10pH
  • Reduced need for mobile-phase modifiers
  • Long column life
  • C18 bonding is monomeric

Specifications

Diameter (Metric) 4.6 mm
For Use With Analysis of a Wide Range of Analytes and Impressive Peak Symmetry for Both Acidic and Basic Compounds
Length (Metric) 100 mm
Particle Size 4 μm
Column Type C18
Type Column
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.