missing translation for 'onlineSavingsMsg'
Learn More

GLYR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen GLYR1
Concentration 0.35 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803694
View Documents
Proteintech
14833-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16893684
View Documents
Proteintech
14833-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated ′Lys-36′ of histone H3 (H3K36me3).
Specifications

Specifications

GLYR1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Rat, Human, Mouse
Q49A26, Q5RKH0, Q922P9
360477, 74022, 84656
GLYR1 Fusion Protein Ag6637
Primary
-20°C
GLYR1
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
BM045, Glyoxylate reductase 1 homolog, GLYR1, HIBDL, N PAC, N-PAC, Nuclear protein NP60, Nuclear protein of 60 kDa, Putative oxidoreductase GLYR1
Glyr1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.