missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLYR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14833-1-AP-20UL
This item is not returnable.
View return policy
Description
GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated ′Lys-36′ of histone H3 (H3K36me3).Specifications
| GLYR1 | |
| Polyclonal | |
| Unconjugated | |
| GLYR1 | |
| BM045, Glyoxylate reductase 1 homolog, GLYR1, HIBDL, N PAC, N-PAC, Nuclear protein NP60, Nuclear protein of 60 kDa, Putative oxidoreductase GLYR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360477, 74022, 84656 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q49A26, Q5RKH0, Q922P9 | |
| Glyr1 | |
| GLYR1 Fusion Protein Ag6637 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction