Learn More
GLS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29519-1-AP-20UL
 View more versions of this product
Product Code. 17561416
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The GLS antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human GLS. This antibody recognizes human antigen. The GLS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GLS
Polyclonal
Rabbit
Primary
Human
Western Blot
Unconjugated
20 μL
2744