Learn More
GLS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29519-1-AP-150UL
Description
The GLS antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human GLS. This antibody recognizes human antigen. The GLS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GLS | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot | |
| Unconjugated | |
| 150 μL | |
| 2744 | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.