Learn More
EKASTU Safety Gas Filter DIRIN 230
EKASTU Gas Filter DIRIN 230 offers protection against harmful gases and vapors
Brand:  EKASTU Safety 422762
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10047900
  • 26.34 € / Each
Estimated Shipment: 21-07-2023
to see stock.
Add to basket
Specifications
DIN EN 14387
Black
Case: Plastic (ABS); Sorbents: Impregnated active charcoal
Protection against inorganic gases and vapors
DIRIN 230 B2
Round thread connection (DIN EN 148-1)
No
Gas Filter
Full masks (DIN EN 136) and half masks (DINEN 140)
10 x 10 x 10 cm