missing translation for 'onlineSavingsMsg'
Learn More

Gallium solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™

Catalog Number p-7115361
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 100 mL
Solvent or Matrix HNO3
Chemical Name or Material Gallium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'