missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH FORTUNA™ Dr. Schilling Pattern Automatic Burette with Amber Glass

Obtain better accuracy and repeatability while simultaneously reducing sample usage. Poulten & Graf GmbH FORTUNA™ Dr. Schilling pattern Automatic Burette with Amber Glass removes the subjective element seen with manual titration.

Brand:  Poulten & Graf GmbH 1 06932

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10177621

  • 114.00 € / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Automatic zero adjustment
  • Dr. Schilling pattern
  • Amber soda lime glass
  • White graduation
  • Schellbach
Specifications

Specifications

5 mL
0.05 mL
White
Schilling Buret
Soda Lime Glass
Plastic Foot, Polyethylene Bottle
0.02 mL
Class B
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions