Learn More
Folic Acid Hydrate EP Impurity B (as Sulfate), Mikromol™
Description
Folic Acid Hydrate EP Impurity B (as Sulfate)
Specifications
Specifications
| Chemical Name or Material | 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate |
| CAS | 39267-74-8 |
| Analyte or Component Names | 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H7 N5 O . H2 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2,5,6-Triaminopyrimidin-4(1H)-one Sulphate, Folic Acid Imp. B (EP) as Sulphate, Folic Acid Hydrate Impurity B as Sulfate, Folic Acid Impurity B as Sulfate |
| InChI Formula | InChI=1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4) |
| SMILES | NC1=NC(=O)C(=C(N)N1)N.OS(=O)(=O)O |
| IUPAC Name | sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one |
| Show More |