Learn More
Fluphenazine N1-Oxide Dihydrochloride, Mikromol™
Description
Fluphenazine N1-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Fluphenazine N1-Oxide Dihydrochloride |
| CAS | 2724689-70-5 |
| Analyte or Component Names | Fluphenazine N1-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H26 F3 N3 O2 S . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, 1-oxide, dihydrochloride (1:2), Fluphenazine N1-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C22H26F3N3O2S.2ClH/c23-22(24,25)17-6-7-21-19(16-17)27(18-4-1-2-5-20(18)31-21)9-3-8-26-10-12-28(30,13-11-26)14-15-29;;/h1-2,4-7,16,29H,3,8-15H2;2*1H |
| SMILES | Cl.Cl.OCC[N+]1([O-])CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 2-[1-oxido-4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-ium-1-yl]ethanol;dihydrochloride |
| Show More |