Learn More
Flupentixol N1-Oxide, Mikromol™
Description
Flupentixol N1-Oxide
Specifications
Specifications
| Chemical Name or Material | Flupentixol N1-Oxide |
| CAS | 2734534-34-8 |
| Analyte or Component Names | Flupentixol N1-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H25 F3 N2 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-[3-[2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]-1-piperazineethanol 1-oxide, 9H-Thioxanthene, 1-piperazineethanol deriv., Flupentixol N-oxide, Flupentixol N1-Oxide |
| InChI Formula | InChI=1S/C23H25F3N2O2S/c24-23(25,26)17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)31-22)5-3-9-27-10-12-28(30,13-11-27)14-15-29/h1-2,4-8,16,29H,3,9-15H2 |
| SMILES | OCC[N+]1([O-])CCN(CCC=C2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 2-[1-oxido-4-[3-[2-(trifluoromethyl)thioxanthen-9-ylidene]propyl]piperazin-1-ium-1-yl]ethanol |
| Show More |