Learn More
Cytiva Fluoro Rubber Peristaltic Tubing
Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3.1 mm
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With | Peristaltic Pump P-1 |
| Type | Peristaltic Tubing |
| Diameter (Metric) Inner | 2.1 mm |
| Overall Length (Metric) | 140 mm |
| Material | Fluoro Rubber |