Learn More
Flunixin Isopropyl Ester, Mikromol™
Description
Flunixin Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Flunixin Isopropyl Ester |
| CAS | 2731709-45-6 |
| Analyte or Component Names | Flunixin Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 F3 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Flunixin Isopropyl Ester |
| InChI Formula | InChI=1S/C17H17F3N2O2/c1-10(2)24-16(23)12-6-5-9-21-15(12)22-14-8-4-7-13(11(14)3)17(18,19)20/h4-10H,1-3H3,(H,21,22) |
| SMILES | CC(C)OC(=O)c1cccnc1Nc2cccc(c2C)C(F)(F)F |
| IUPAC Name | propan-2-yl 2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylate |
| Show More |