Learn More
Flunitrazepam EP Impurity B, Mikromol™
Product Code. 17962596
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962596 100 mg 100mg
17913226 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17962596 Supplier Mikromol Supplier No. MM0069.02
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Flunitrazepam EP Impurity B
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desmethylflunitrazepam
CAS 2558-30-7
Analyte or Component Names Desmethylflunitrazepam
Shipping Condition Room Temperature
Molecular Formula C15 H10 F N3 O3
Certifications/Compliance ISO 17025
Synonym 5-(2-Fluorophenyl)-7-nitro-1,3-dihydro-2H-1,4-benzodiazepin-2-one (Demethylflunitrazepam), 5-(2-Fluorophenyl)-7-nitro-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Flunitrazepam Imp. B (EP), Demethylflunitrazepam, Desmethylflunitrazepam, Flunitrazepam Impurity B
InChI Formula InChI=1S/C15H10FN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)
SMILES [O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3F)c2c1
IUPAC Name 5-(2-fluorophenyl)-7-nitro-1,3-dihydro-1,4-benzodiazepin-2-one
Molecular Weight (g/mol) 299.26
Quantity 100 mg
Formula Weight 299.0706 g/mol