Learn More
Flufenoximacil, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Flufenoximacil |
| CAS | 2759011-88-4 |
| Analyte or Component Names | Flufenoximacil |
| Shipping Condition | +20°C |
| Molecular Formula | C17H14ClF4N3O5 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Methyl (2R)-2-[[(E)-[[2-chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluorophenyl]methylene]amino]oxy]propanoate |
| InChI Formula | InChI=1S/C17H14ClF4N3O5/c1-8(15(27)29-3)30-23-7-9-4-12(11(19)5-10(9)18)25-14(26)6-13(17(20,21)22)24(2)16(25)28/h4-8H,1-3H3/b23-7+/t8-/m1/s1 |
| SMILES | O=C(N1C)N(C(C(F)=C2)=CC(/C=N/O[C@H](C)C(OC)=O)=C2Cl)C(C=C1C(F)(F)F)=O |
| IUPAC Name | methyl (2R)-2-[(E)-[2-chloro-4-fluoro-5-[3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]phenyl]methylideneamino]oxypropanoate |
| Show More |