Learn More
Flufenamic Acid Methyl Ester, Mikromol™
Description
Flufenamic Acid Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Flufenamic Acid Methyl Ester |
| CAS | 2765-91-5 |
| Analyte or Component Names | Flufenamic Acid Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 F3 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Flufenamic Acid Methyl Ester, 2-[[3-(trifluoromethyl)phenyl]amino]benzoic acid methyl ester, Anthranilic acid, N-(alpha,alpha,alpha-trifluoro-m-tolyl)-, methyl ester (7CI,8CI), Flufenamic acid methyl ester, Methyl flufenamate |
| InChI Formula | InChI=1S/C15H12F3NO2/c1-21-14(20)12-7-2-3-8-13(12)19-11-6-4-5-10(9-11)15(16,17)18/h2-9,19H,1H3 |
| SMILES | COC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | methyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |