Learn More
Flufenamic Acid Ethyl Ester, Mikromol™
Description
Flufenamic Acid Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Flufenamic Acid Ethyl Ester |
| CAS | 2766-16-7 |
| Analyte or Component Names | Flufenamic Acid Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 F3 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Flufenamic Acid Ethyl Ester, 2-[[3-(trifluoromethyl)phenyl]amino]benzoic acid ethyl ester, Anthranilic acid, N-(alpha,alpha,alpha-trifluoro-m-tolyl)-, ethyl ester (7CI,8CI) |
| InChI Formula | InChI=1S/C16H14F3NO2/c1-2-22-15(21)13-8-3-4-9-14(13)20-12-7-5-6-11(10-12)16(17,18)19/h3-10,20H,2H2,1H3 |
| SMILES | CCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | ethyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |