Learn More
Fluensulfone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30324999
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324999 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324999 Supplier Dr. Ehrenstorfer Supplier No. DREA13710900AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fluensulfone
CAS 318290-98-1
Analyte or Component Names Fluensulfone
Shipping Condition Room Temperature
Molecular Formula C7 H5 Cl F3 N O2 S2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Thiazole, 5-chloro-2-[(3,4,4-trifluoro-3-butenyl)sulfonyl]- (9CI), 5-Chloro-2-[(3,4,4-trifluorobut-3-en-1-yl)sulfonyl]-1,3-thiazole, Fluensulfone, MCW 2, Thiazole, 5-chloro-2-[(3,4,4-trifluoro-3-buten-1-yl)sulfonyl]-
InChI Formula InChI=1S/C7H5ClF3NO2S2/c8-5-3-12-7(15-5)16(13,14)2-1-4(9)6(10)11/h3H,1-2H2
SMILES FC(=C(F)CCS(=O)(=O)c1ncc(Cl)s1)F
IUPAC Name 5-chloro-2-(3,4,4-trifluorobut-3-enylsulfonyl)-1,3-thiazole
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 291.7
Quantity 1 mL
Formula Weight 290.9402 g/mol