Learn More
Flucarbazone sodium 100 μg/mL in Acetonitrile/Water, Dr. Ehrenstorfer
Product Code. 30328195
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328195 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328195 Supplier Dr. Ehrenstorfer Supplier No. DREA13685000WL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Flucarbazone-sodium
CAS 181274-17-9
Analyte or Component Names Flucarbazone-sodium
Shipping Condition Room Temperature
Molecular Formula C12 H10 F3 N4 O6 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C12H11F3N4O6S.Na/c1-18-10(24-2)16-19(11(18)21)9(20)17-26(22,23)8-6-4-3-5-7(8)25-12(13,14)15;/h3-6H,1-2H3,(H,17,20);/q;+1/p-1
SMILES [Na+].COC1=NN(C(=O)[N-]S(=O)(=O)c2ccccc2OC(F)(F)F)C(=O)N1C
IUPAC Name sodium;(3-methoxy-4-methyl-5-oxo-1,2,4-triazole-1-carbonyl)-[2-(trifluoromethoxy)phenyl]sulfonylazanide
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 418.28
Quantity 1 mL
Formula Weight 418.0171 g/mol