Learn More
Flavoxate N-Oxide Hydrochloride, Mikromol™
Description
Flavoxate N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Flavoxate N-Oxide Hydrochloride |
| CAS | 2732347-10-1 |
| Analyte or Component Names | Flavoxate N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H25 N O5 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Flavoxate N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C24H25NO5.ClH/c1-17-21(26)19-11-8-12-20(23(19)30-22(17)18-9-4-2-5-10-18)24(27)29-16-15-25(28)13-6-3-7-14-25;/h2,4-5,8-12H,3,6-7,13-16H2,1H3;1H |
| SMILES | Cl.CC1=C(Oc2c(cccc2C1=O)C(=O)OCC[N+]3([O-])CCCCC3)c4ccccc4 |
| IUPAC Name | 2-(1-oxidopiperidin-1-ium-1-yl)ethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate;hydrochloride |
| Show More |