Learn More
Flash Point Standard, COC ASTM D92 (Nominal Value: 85.0 °C), Paragon Scientific™
Click to view available options
Quantity:
3 x 80 mL
Unit Size:
Pack of 3
Description
- Certified Reference Material for annual verification checks in accordance with ASTM D92.
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 3 x 80 mL |
Nominal values quoted. Certified Values will vary from batch to batch.