Learn More
Fisherbrand™ Maxima™ C Plus Pumps, Standard Exhaust Oil Filter Replacement Elements
Replacement filter will help keep the Macima C vacuum pump running smoothly
146.00 € - 254.00 €
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
12905134
|
Fisherbrand™
|
Model M12C and M16C vacuum pump exhaust filter |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10060011
|
Fisherbrand™
|
Model M2C vacuum pump exhaust filter |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Replacement filter will help keep the Macima C vacuum pump running smoothly
For exhaust filters supplied with Fisher Scientific Maxima C Plus pumps.
Specifications
Filter Element |