Learn More
Finasteride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Finasteride
Specifications
Specifications
| Chemical Name or Material | Finasteride |
| CAS | 98319-26-7 |
| Analyte or Component Names | Finasteride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H36 N2 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Finasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxamide, N-(1,1-dimethylethyl)-3-oxo-, (5alpha,17beta)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-(1,1-Dimethylethyl)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, Appecia, Chibro-Proscar, Finalo, Finara, Finast, Finasterid Alternova, Finastid, Finax, Fincar, Finpecia, Fistide, Geffina, MK 906, N-(tert-Butyl)-3-oxo-4-aza-5alpha-androst-1-ene-17-carboxamide, Propecia, Proscar, Prosteride, Prostide |
| InChI Formula | InChI=1S/C23H36N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h11,13-18H,6-10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1 |
| SMILES | CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| Show More |