Learn More
Fenbufen Isopropyl Ester, Mikromol™
Product Code. 30051603
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051603 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051603 Supplier Mikromol Supplier No. MM0918.080025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fenbufen Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fenbufen Isopropyl Ester
CAS 2725579-21-3
Analyte or Component Names Fenbufen Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C19 H20 O3
Certifications/Compliance ISO 9001
Synonym Fenbufen Isopropyl Ester
InChI Formula InChI=1S/C19H20O3/c1-14(2)22-19(21)13-12-18(20)17-10-8-16(9-11-17)15-6-4-3-5-7-15/h3-11,14H,12-13H2,1-2H3
SMILES CC(C)OC(=O)CCC(=O)c1ccc(cc1)c2ccccc2
IUPAC Name propan-2-yl 4-oxo-4-(4-phenylphenyl)butanoate
Molecular Weight (g/mol) 296.36
Quantity 25 mg
Formula Weight 296.1412 g/mol