missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
213.00 € - 460.00 €
Specifications
| Antigen | FANCG |
|---|---|
| Dilution | Western Blot 1:1000, Immunoblotting reported in scientific literature (PMID 24910428) |
| Applications | Western Blot, Immunoblot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18048633
|
Novus Biologicals
NB100-2566SS |
0.025 mL |
213.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18739273
|
Novus Biologicals
NB100-2566 |
0.1 mL |
460.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCG Polyclonal specifically detects FANCG in Human samples. It is validated for Western Blot, Immunoblotting.Specifications
| FANCG | |
| Western Blot, Immunoblot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| Whole antisera with No Preservative | |
| DNA repair protein XRCC9, FAG, Fanconi anemia group G protein, Fanconi anemia, complementation group G, Protein FACG, X-ray repair, complementing defective, in Chinese hamster, 9, XRCC9X-ray repair complementing defective repair in Chinese hamster cells 9 | |
| FANCG | |
| IgG | |
| Unpurified | |
| Human. Immunogen sequence has 88% homology to mouse. |
| Western Blot 1:1000, Immunoblotting reported in scientific literature (PMID 24910428) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| O15287 | |
| 2189 | |
| A peptide corresponding to the C-terminus of human FANCG (within amino acids 550-622). [SwissProt# O15287] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 69 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title