Learn More
FANCG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NB100-2566
 View more versions of this product
Product Code. 18739273
  • 460.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
FANCG Polyclonal specifically detects FANCG in Human samples. It is validated for Western Blot, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Rabbit
0.1 mL
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Whole antisera with No Preservative
69 kDa
RUO
Antisera